Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNMA1 SIRNA Assays ›
Gene Symbol:
KCNMA1
Gene Name:
potassium calcium-activated channel subfamily M alpha 1
Gene Aliases:
BKTM, KCa1.1, MaxiK, SAKCA, SLO, SLO-ALPHA, SLO1, bA205K10.1, hSlo, mSLO1
Chromosome Location:
Chr.10: 76869601 - 77637819 on Build GRCh38
UniGene ID:
Hs.144795
Species:
Human
siRNA ID
HSS180152
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNMA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001014797.2
NP_001014797.1
21, 22
2497
NM_001161352.1
NP_001154824.1
21, 22
2659
NM_001271518.1
NP_001258447.1
20, 21
2335
NM_001271519.1
NP_001258448.1
21, 22
2494
NM_001322829.1
NP_001309758.1
21, 22
2494
NM_001322830.1
NP_001309759.1
22, 23
2506
NM_001322832.1
NP_001309761.1
20, 21
2485
NM_001322835.1
NP_001309764.1
21, 22
2497
NM_001322836.1
NP_001309765.1
21, 22
2494
NM_001322837.1
NP_001309766.1
21, 22
2497
NM_001322838.1
NP_001309767.1
22, 23
2532
NM_002247.3
NP_002238.2
20, 21
2485
XM_005269776.3
XP_005269833.1
20, 21
2474
XM_005269778.1
XP_005269835.1
22, 23
2660
XM_005269781.2
XP_005269838.1
21, 22
2580
XM_005269787.3
XP_005269844.1
20, 21
2474
XM_005269789.2
XP_005269846.1
21, 22
2418
XM_005269792.2
XP_005269849.1
21, 22
2493
XM_005269796.2
XP_005269853.1
20, 21
2406
XM_006717826.2
XP_006717889.1
20, 21
2406
XM_011539773.1
XP_011538075.1
23, 24
2792
XM_011539774.1
XP_011538076.1
23, 24
2705
XM_011539775.1
XP_011538077.1
22, 23
2780
XM_011539776.1
XP_011538078.1
22, 23
2780
XM_011539777.1
XP_011538079.1
22, 23
2660
XM_011539778.2
XP_011538080.1
21, 22
2580
XM_011539779.1
XP_011538081.1
22, 23
2693
XM_011539780.1
XP_011538082.1
22, 23
2660
XM_011539781.2
XP_011538083.1
22, 23
2618
XM_011539782.2
XP_011538084.1
21, 22
2558
XM_011539783.1
XP_011538085.1
22, 23
2573
XM_011539785.1
XP_011538087.1
21, 22
2483
XM_017016207.1
XP_016871696.1
23, 24
2792
XM_017016208.1
XP_016871697.1
23, 24
2792
XM_017016209.1
XP_016871698.1
22, 23
2780
XM_017016210.1
XP_016871699.1
23, 24
2792
XM_017016211.1
XP_016871700.1
23, 24
2705
XM_017016212.1
XP_016871701.1
22, 23
2618
XM_017016213.1
XP_016871702.1
22, 23
2693
XM_017016214.1
XP_016871703.1
21, 22
2606
XM_017016215.1
XP_016871704.1
22, 23
2573
XM_017016216.1
XP_016871705.1
22, 23
2618
XM_017016217.1
XP_016871706.1
21, 22
2558
XM_017016218.1
XP_016871707.1
20, 21
1943
XM_017016222.1
XP_016871711.1
20, 21
2474
XM_017016223.1
XP_016871712.1
19, 20
1870
GenBank mRNA
AB113382.1
-
21, 22
2424
AB209488.1
-
20, 21
1904
AB524033.1
-
20, 21
2158
AF025999.1
-
20, 21
2236
AF118141.1
-
20, 21
2113
AK310379.1
-
22, 23
2532
AY040849.1
-
20, 21
2250
BC137115.1
-
20, 21
2388
9021112
BC137137.1
-
20, 21
2388
9021134
BC144496.1
-
21, 22
2397
9053023
BX648925.1
-
17, 18
1650
U02632.1
-
14, 15
1430
U11058.2
-
20, 21
2199
U11717.1
-
20, 21
2260
U13913.1
-
20, 21
2386
U23767.1
-
20, 21
2308
Back To Top
More Information
Panther Classification:
Molecular Function -
ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
potassium ion transport
response to osmotic stress
cellular potassium ion homeostasis
response to carbon monoxide
regulation of ion transmembrane transport
regulation of membrane potential
positive regulation of apoptotic process
negative regulation of cell volume
response to calcium ion
micturition
smooth muscle contraction involved in micturition
potassium ion transmembrane transport
actin binding
voltage-gated potassium channel activity
protein binding
calcium-activated potassium channel activity
metal ion binding
large conductance calcium-activated potassium channel activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0