Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207370.2 | NP_997253.2 | 3 | 1031 | ||
XM_011541434.2 | XP_011539736.1 | 3 | 1135 | |||
XM_017001250.1 | XP_016856739.1 | 2 | 1117 | |||
XM_017001251.1 | XP_016856740.1 | 3 | 1020 | |||
GenBank mRNA | AY255529.1 | - | 1 | 294 | ||
BC068275.1 | - | 3 | 1031 | 5167285 |