Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001732.3 | NP_001001732.2 | 5 | 533 | ||
NM_001159747.1 | NP_001153219.1 | 6 | 566 | |||
XM_011539789.2 | XP_011538091.1 | 13 | 1627 | |||
XM_011539790.2 | XP_011538092.1 | 13 | 1627 | |||
XM_011539791.2 | XP_011538093.1 | 12 | 1444 | |||
XM_011539792.2 | XP_011538094.1 | 7 | 749 | |||
XM_011539793.2 | XP_011538095.1 | 6 | 1728 | |||
XM_011539795.2 | XP_011538097.1 | 5 | 1545 | |||
XM_017016240.1 | XP_016871729.1 | 6 | 805 | |||
GenBank mRNA | AK302377.1 | - | 6 | 545 | ||
BC075861.1 | - | 5 | 498 | 5171414 |