Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008395.3 | NP_001008396.1 | 2 | 122 | ||
NM_001318236.1 | NP_001305165.1 | 2 | 122 | |||
NR_134526.1 | 2 | 122 | ||||
NR_134527.1 | 2 | 122 | ||||
NR_134528.1 | 2 | 121 | ||||
NR_134529.1 | 2 | 101 | ||||
NR_134530.1 | 2 | 121 | ||||
XM_017012198.1 | XP_016867687.1 | 4 | 509 | |||
XM_017012199.1 | XP_016867688.1 | 4 | 509 | |||
GenBank mRNA | AA419348.1 | - | 2 | 97 | ||
AJ714519.1 | - | 2 | 102 | |||
BC063401.1 | - | 2 | 102 | 5194116 | ||
BC105609.1 | - | 2 | 112 | 30321610 | ||
BC130553.1 | - | 2 | 69 | 40146584 | ||
BC130559.1 | - | 2 | 69 | 40146590 | ||
BE733059.1 | - | 1 | 32 | |||
BF978089.1 | - | 121 | ||||
BG706819.1 | - | 2 | 105 | |||
BG715626.1 | - | 2 | 119 | |||
BG819624.1 | - | 1 | 40 | |||
BI598677.1 | - | 2 | 125 | |||
CU450474.1 | - | 2 | 175 | |||
DB498044.1 | - | 2 | 100 | |||
DB539334.2 | - | 1 | 58 |