Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KTN1 SIRNA Assays ›
Gene Symbol:
KTN1
Gene Name:
kinectin 1
Gene Aliases:
CG1, KNT, MU-RMS-40.19
Chromosome Location:
Chr.14: 55580207 - 55684584 on Build GRCh38
UniGene ID:
Hs.509414
Species:
Human
siRNA ID
HSS180422
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001079521.1
NP_001072989.1
26, 27
2866
NM_001079522.1
NP_001072990.1
26, 27
2921
NM_001271014.1
NP_001257943.1
27, 28
2990
NM_004986.3
NP_004977.2
26, 27
2866
NR_073128.1
26, 27
2921
NR_073129.1
27, 28
2990
XM_006720138.2
XP_006720201.1
27, 28
2981
XM_006720139.2
XP_006720202.1
26, 27
2829
XM_006720140.3
XP_006720203.1
26, 27
2829
XM_006720141.2
XP_006720204.1
26, 27
2829
XM_011536751.2
XP_011535053.1
26, 27
3087
XM_011536752.1
XP_011535054.1
25, 26
2760
XM_011536753.1
XP_011535055.1
25, 26
2760
XM_017021278.1
XP_016876767.1
26, 27
8679
XM_017021279.1
XP_016876768.1
25, 26
2760
XM_017021280.1
XP_016876769.1
26, 27
2912
XM_017021281.1
XP_016876770.1
26, 27
8734
XM_017021282.1
XP_016876771.1
27, 28
2984
XM_017021283.1
XP_016876772.1
27, 28
2918
XM_017021284.1
XP_016876773.1
25, 26
2760
XM_017021285.1
XP_016876774.1
26, 27
2892
XM_017021286.1
XP_016876775.1
25, 26
2760
GenBank mRNA
AK302797.1
-
27, 28
2944
AK308272.1
-
9, 10
686
AY264265.1
-
27, 28
2875
AY536375.1
-
16, 17
1079
BC112337.1
-
26, 27
2804
40068009
BC117132.1
-
25, 26
2738
40125683
BC143720.1
-
26, 27
2807
9052234
D13629.1
-
26, 27
2771
L25616.1
-
25, 26
2728
Z22551.1
-
26, 27
2754
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
cell-cell adhesion
protein binding
kinesin binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0