Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTRHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013663.1 | NP_001013685.1 | 2 | 346 | ||
GenBank mRNA | BC073803.1 | - | 2 | 346 | 4896064 |