Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193482.1 | NP_001180411.1 | 10 | 1002 | ||
NM_001193483.2 | NP_001180412.1 | 10 | 1147 | |||
NM_001193484.1 | NP_001180413.1 | 10 | 1123 | |||
NM_001193485.2 | NP_001180414.1 | 10 | 1352 | |||
NM_001193488.1 | NP_001180417.1 | 10 | 1070 | |||
NM_004987.5 | NP_004978.2 | 10 | 1032 | |||
XM_005263946.1 | XP_005264003.1 | 11 | 1344 | |||
XM_005263948.3 | XP_005264005.1 | 11 | 1397 | |||
XM_005263949.1 | XP_005264006.1 | 11 | 1339 | |||
XM_005263950.1 | XP_005264007.1 | 11 | 1180 | |||
XM_005263951.4 | XP_005264008.1 | 11 | 1616 | |||
XM_017004091.1 | XP_016859580.1 | 11 | 1366 | |||
XM_017004092.1 | XP_016859581.1 | 11 | 1361 | |||
XM_017004093.1 | XP_016859582.1 | 11 | 1410 | |||
XM_017004094.1 | XP_016859583.1 | 11 | 1645 | |||
GenBank mRNA | AF527759.1 | - | 10 | 1352 | ||
AF527760.1 | - | 10 | 1104 | |||
AF527761.1 | - | 10 | 1080 | |||
AF527762.1 | - | 10 | 1013 | |||
AF527763.1 | - | 10 | 1002 | |||
AK296992.1 | - | 10 | 1102 | |||
AK302411.1 | - | 10 | 1127 | |||
AK304260.1 | - | 10 | 1123 | |||
AK314217.1 | - | 10 | 1032 | |||
BC005341.1 | - | 10 | 1070 | 3838632 | ||
U09284.2 | - | 10 | 1022 |