Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320870.1 | NP_001307799.1 | 2 | 955 | ||
NM_001320871.1 | NP_001307800.1 | 2 | 955 | |||
NM_001320872.1 | NP_001307801.1 | 2 | 955 | |||
NM_201520.2 | NP_958928.1 | 2 | 955 | |||
NR_135483.1 | 2 | 955 | ||||
NR_135484.1 | 2 | 955 | ||||
XM_005256641.2 | XP_005256698.1 | 2 | 1012 | |||
XM_011523846.1 | XP_011522148.1 | 2 | 1012 | |||
XM_017024633.1 | XP_016880122.1 | 2 | 1012 | |||
XM_017024634.1 | XP_016880123.1 | 2 | 1306 | |||
GenBank mRNA | AK097536.1 | - | 2 | 464 | ||
AY498866.1 | - | 2 | 419 | |||
BC041597.1 | - | 2 | 955 | |||
BC052233.1 | - | 2 | 955 | |||
BC101329.2 | - | 2 | 450 | 40025001 | ||
BC101330.2 | - | 2 | 448 | 40025003 | ||
BC105995.1 | - | 2 | 930 | 4746077 |