Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013672.4 | NP_001013694.4 | 2 | 1265 | ||
GenBank mRNA | AK128660.1 | - | 1355 | |||
BC057385.1 | - | 2 | 1246 | |||
BC070116.1 | - | 2 | 1191 | |||
BC141806.1 | - | 3 | 1264 | 40147362 | ||
KR780251.1 | - | 2 | 1265 |