Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136536.4 | NP_001130008.2 | 7 | 1714 | ||
NM_001170700.2 | NP_001164171.1 | 7 | 1869 | |||
XM_006714014.3 | XP_006714077.1 | 8 | 2348 | |||
XM_011513693.2 | XP_011511995.1 | 9 | 2376 | |||
XM_011513694.2 | XP_011511996.1 | 9 | 2313 | |||
XM_011513695.2 | XP_011511997.1 | 7 | 2186 | |||
XM_011513696.2 | XP_011511998.1 | 8 | 1759 | |||
XM_017008191.1 | XP_016863680.1 | 9 | 2376 | |||
XM_017008192.1 | XP_016863681.1 | 9 | 2378 | |||
GenBank mRNA | AK131492.1 | - | 7 | 1707 | ||
AK304357.1 | - | 7 | 1700 | |||
AK316523.1 | - | 7 | 1700 | |||
BC157885.1 | - | 7 | 1869 | 9057096 |