Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114726.2 | NP_001108198.2 | 6 | 1568 | ||
NM_001174164.1 | NP_001167635.1 | 6 | 2719 | |||
XM_005250342.3 | XP_005250399.1 | 5 | 2719 | |||
XM_005250343.3 | XP_005250400.1 | 5 | 2628 | |||
XM_006715985.3 | XP_006716048.1 | 5 | 2732 | |||
XM_017012205.1 | XP_016867694.1 | 6 | 3538 | |||
XM_017012206.1 | XP_016867695.1 | 6 | 2477 |