Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166400.1 | NP_001159872.1 | 4 | 1355 | ||
NM_001166401.1 | NP_001159873.1 | 3 | 1327 | |||
NM_005364.4 | NP_005355.2 | 3 | 1076 | |||
XM_017029520.1 | XP_016885009.1 | 4 | 1143 | |||
GenBank mRNA | AK313943.1 | - | 3 | 1076 | ||
BC002455.2 | - | 3 | 1022 | 3345962 | ||
BC012744.2 | - | 3 | 1015 | 3617812 | ||
BT007340.1 | - | 1 | 806 |