Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166343.1 | NP_001159815.1 | 2 | 485 | ||
NM_002405.3 | NP_002396.2 | 3 | 580 | |||
NR_029413.1 | 2 | 531 | ||||
GenBank mRNA | AK292123.1 | - | 3 | 521 | ||
AK297149.1 | - | 2 | 479 | |||
AK297833.1 | - | 2 | 464 | |||
AK316555.1 | - | 2 | 472 | |||
BC094814.1 | - | 3 | 541 | 5930073 | ||
BM457814.1 | - | 3 | 535 | |||
CR456518.1 | - | 3 | 373 | |||
U94352.1 | - | 3 | 487 |