Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CTAGE5 SIRNA Assays ›
Gene Symbol:
CTAGE5
Gene Name:
CTAGE family member 5, ER export factor
Gene Aliases:
MEA6, MGEA, MGEA11, MGEA6
Chromosome Location:
Chr.14: 39265264 - 39351193 on Build GRCh38
UniGene ID:
Hs.509200
Species:
Human
siRNA ID
HSS181098
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CTAGE5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001247988.1
NP_001234917.1
2
269
NM_001247989.1
NP_001234918.1
2
438
NM_005930.3
NP_005921.2
2
438
NM_203354.2
NP_976229.1
2
189
NM_203355.2
NP_976230.1
2
438
NM_203356.2
NP_976231.1
2
269
XM_005267648.4
XP_005267705.1
2
149
XM_006720148.2
XP_006720211.1
2
151
XM_011536775.2
XP_011535077.1
2
489
XM_011536778.2
XP_011535080.1
2
177
XM_017021314.1
XP_016876803.1
2
490
XM_017021315.1
XP_016876804.1
2
191
XM_017021316.1
XP_016876805.1
2
178
XM_017021317.1
XP_016876806.1
2
599
XM_017021318.1
XP_016876807.1
2
220
XM_017021319.1
XP_016876808.1
2
156
XM_017021320.1
XP_016876809.1
2
224
XM_017021321.1
XP_016876810.1
2
490
XM_017021322.1
XP_016876811.1
2
197
XM_017021325.1
XP_016876814.1
2
157
XM_017021330.1
XP_016876819.1
2
99
GenBank mRNA
AB209528.1
-
1
747
AF338233.1
-
2
283
AF338234.1
-
2
316
AK055791.1
-
2
558
AK091252.1
-
2
647
AK298935.1
-
2
170
AK315254.1
-
2
159
AL832233.1
-
2
152
AL833415.1
-
2
98
BC029513.1
-
2
156
BC039017.1
-
2
195
BC064355.1
-
2
384
4803802
BX367676.2
-
2
388
BX640994.1
-
2
170
BX648014.1
-
2
170
CD512382.1
-
2
176
CV809109.1
-
2
460
DA072098.1
-
2
189
U73682.1
-
2
148
U94780.1
-
2
416
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
Gene Ontology Categories:
Function(s)
Process(es)
ER to Golgi vesicle-mediated transport
protein exit from endoplasmic reticulum
cargo loading into vesicle
positive regulation of catalytic activity
protein localization to endoplasmic reticulum exit site
regulation of guanyl-nucleotide exchange factor activity
protein binding
enzyme activator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0