Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029954.2 | NP_001025125.2 | 3 | 374 | ||
XM_011519488.2 | XP_011517790.1 | 3 | 463 | |||
GenBank mRNA | BC037872.1 | - | 5 | 539 | ||
BC133042.1 | - | 3 | 361 | 40147064 | ||
BC133044.1 | - | 3 | 361 | 40147066 | ||
BC144619.1 | - | 3 | 361 | |||
CN311321.1 | - | 3 | 350 |