Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RERE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042681.1 | NP_001036146.1 | 18 | 2657 | ||
NM_001042682.1 | NP_001036147.1 | 8 | 956 | |||
NM_012102.3 | NP_036234.3 | 19 | 2842 | |||
XM_005263464.2 | XP_005263521.1 | 18 | 2596 | |||
XM_005263466.1 | XP_005263523.1 | 12 | 1479 | |||
XM_011541510.1 | XP_011539812.1 | 17 | 2582 | |||
XM_011541511.1 | XP_011539813.1 | 18 | 2708 | |||
XM_017001358.1 | XP_016856847.1 | 18 | 2347 | |||
XM_017001359.1 | XP_016856848.1 | 18 | 2852 | |||
GenBank mRNA | AB007927.2 | - | 17 | 1992 | ||
AB036737.1 | - | 19 | 2668 | |||
AF016005.1 | - | 8 | 934 | |||
AF118275.1 | - | 4 | 370 |