Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 13 | 1569 | ||
NM_002498.2 | NP_002489.1 | 14 | 1620 | |||
NM_152720.2 | NP_689933.1 | 14 | 1639 | |||
NR_027415.2 | 14 | 1619 | ||||
GenBank mRNA | AB072828.1 | - | 12 | 1238 | ||
AK131359.1 | - | 12 | 2487 | |||
AK290259.1 | - | 14 | 1352 | |||
AL833951.1 | - | 13 | 5002 | |||
AW364762.1 | - | 4 | 351 | |||
BC019916.1 | - | 14 | 1347 | 5000918 | ||
BX395059.2 | - | 3 | 735 | |||
Z29067.1 | - | 12 | 1103 |