Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146099.1 | NP_001139571.1 | 3 | 564 | ||
NM_002498.2 | NP_002489.1 | 3 | 564 | |||
NM_152720.2 | NP_689933.1 | 3 | 583 | |||
NR_027415.2 | 3 | 564 | ||||
GenBank mRNA | AB072828.1 | - | 2 | 233 | ||
AK131359.1 | - | 2 | 1482 | |||
AK290259.1 | - | 3 | 296 | |||
AL833951.1 | - | 3 | 423 | |||
BC019916.1 | - | 3 | 291 | 5000918 | ||
BQ648157.1 | - | 3 | 500 | |||
CV029372.1 | - | 2 | 188 | |||
DA733733.1 | - | 3 | 564 | |||
Z29067.1 | - | 1 | 47 |