Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004556.2 | NP_004547.2 | 1 | 774 | ||
GenBank mRNA | AK301360.1 | - | 1 | 538 | ||
BC011676.2 | - | 1 | 520 | |||
BC063609.1 | - | 1 | 520 | |||
BX367256.2 | - | 1 | 612 | |||
U91616.1 | - | 1 | 782 |