Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP1A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001734.1 | NP_001001734.1 | 1 | 201 | ||
NM_144699.3 | NP_653300.2 | 18 | 3162 | |||
XM_011509582.1 | XP_011507884.1 | 17 | 2629 | |||
GenBank mRNA | AF506797.1 | - | 18 | 3154 | ||
AK098076.1 | - | 8 | 1039 | |||
BC028297.1 | - | 1 | 201 | |||
BC094801.1 | - | 18 | 3113 | 5268907 | ||
BX647234.1 | - | 18 | 3136 |