Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NINJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294345.1 | NP_001281274.1 | 2 | 211 | ||
NM_001294346.1 | NP_001281275.1 | 2 | 318 | |||
NM_016533.5 | NP_057617.2 | 2 | 557 | |||
GenBank mRNA | AK296012.1 | - | 2 | 557 | ||
AK298043.1 | - | 2 | 269 | |||
AK315995.1 | - | 2 | 555 | |||
BC057766.1 | - | 2 | 225 | |||
BI597990.1 | - | 2 | 410 | |||
BM128205.1 | - | 2 | 179 | |||
CB995632.1 | - | 2 | 238 | |||
CD695373.1 | - | 2 | 305 |