Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007176.4 | NP_001007177.1 | 3 | 268 | ||
NM_001256596.1 | NP_001243525.1 | 3 | 160 | |||
NM_001256597.1 | NP_001243526.1 | 3 | 268 | |||
NM_001256598.1 | NP_001243527.1 | 3 | 268 | |||
GenBank mRNA | AJ276240.1 | - | 3 | 255 | ||
BC017981.1 | - | 3 | 261 | 4245342 | ||
BG202040.1 | - | 3 | 186 | |||
DA820911.1 | - | 3 | 269 | |||
DA900243.1 | - | 3 | 268 |