Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024822.2 | NP_001019993.1 | 1 | 526 | ||
GenBank mRNA | AW274139.1 | - | 1 | 219 | ||
AY665807.1 | - | 1 | 426 | |||
BC130347.1 | - | 1 | 525 | 40146378 | ||
BC130349.1 | - | 1 | 526 | 40146380 | ||
FJ237362.1 | - | 1 | 526 |
Gene Symbol: RNASE11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY358794.1 | - | 1 | 242 |