Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035005.3 | NP_001030177.1 | 3 | 435 | ||
NM_001199346.1 | NP_001186275.1 | 3 | 271 | |||
GenBank mRNA | AB097012.1 | - | 3 | 355 | ||
AK027111.1 | - | 3 | 231 | |||
AK291882.1 | - | 3 | 242 | |||
BC022357.1 | - | 3 | 240 | 4297017 | ||
BC093004.1 | - | 3 | 392 | 5273608 |
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 7 | 723 | ||
NM_001199356.1 | NP_001186285.1 | 7 | 790 | |||
GenBank mRNA | BX393840.2 | - | 7 | 725 |