Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014080.4 | NP_054799.4 | 3 | 319 | ||
XM_005254421.2 | XP_005254478.1 | 3 | 319 | |||
GenBank mRNA | AB209010.1 | - | 2 | 159 | ||
AF230496.1 | - | 3 | 301 | |||
AF267981.1 | - | 3 | 319 | |||
BI045475.1 | - | 3 | 505 |