Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016068.2 | NP_057152.2 | 4 | 358 | ||
GenBank mRNA | AF151893.1 | - | 4 | 353 | ||
AK310968.1 | - | 3 | 512 | |||
AK311665.1 | - | 4 | 626 | |||
BC003540.1 | - | 4 | 336 | 3611166 | ||
BC009428.1 | - | 4 | 336 | 3611056 | ||
BI836529.1 | - | 4 | 358 |