Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127582.1 | NP_001121054.1 | 5 | 526 | ||
NM_001127583.1 | NP_001121055.1 | 5 | 517 | |||
NM_001127584.1 | NP_001121056.1 | 5 | 514 | |||
NM_001127585.1 | NP_001121057.1 | 4 | 454 | |||
NM_001127586.1 | NP_001121058.1 | 5 | 526 | |||
NM_016162.3 | NP_057246.2 | 5 | 523 | |||
XM_005253698.3 | XP_005253755.1 | 6 | 774 | |||
XM_011520964.2 | XP_011519266.1 | 4 | 452 | |||
XM_011520965.2 | XP_011519267.1 | 5 | 473 | |||
XM_017019392.1 | XP_016874881.1 | 6 | 771 | |||
XM_017019393.1 | XP_016874882.1 | 6 | 762 | |||
GenBank mRNA | AB197695.1 | - | 5 | 482 | ||
AB197696.1 | - | 5 | 476 | |||
AB197697.1 | - | 5 | 473 | |||
AF063594.1 | - | 5 | 501 | |||
AF110645.1 | - | 5 | 502 | |||
AF156552.1 | - | 5 | 502 | |||
AK293169.1 | - | 4 | 439 | |||
AK303354.1 | - | 5 | 508 | |||
AK313621.1 | - | 5 | 495 | |||
BC007781.2 | - | 5 | 488 | 4309278 | ||
BC013038.2 | - | 5 | 468 | |||
BC095434.1 | - | 5 | 487 | 30365729 | ||
BG705367.1 | - | 5 | 514 | |||
BG718170.1 | - | 5 | 534 | |||
BM922497.1 | - | 6 | 752 | |||
BP303219.1 | - | 5 | 523 | |||
EF152349.1 | - | 4 | 413 | |||
EF152350.1 | - | 4 | 318 | |||
EF152351.1 | - | 5 | 485 | |||
EF152352.1 | - | 5 | 485 |