Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCOLCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002593.3 | NP_002584.2 | 5 | 848 | ||
GenBank mRNA | AB008549.1 | - | 5 | 706 | ||
AK313748.1 | - | 5 | 744 | |||
BC000574.2 | - | 5 | 741 | 3163857 | ||
BC033205.1 | - | 5 | 686 | 4896776 |