Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016466.5 | NP_057550.3 | 4 | 561 | ||
GenBank mRNA | AF151034.1 | - | 4 | 542 | ||
BC031303.2 | - | 4 | 553 | 5284947 | ||
BU626151.1 | - | 1 | 350 | |||
DA769410.1 | - | 4 | 558 |