Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other UBAP1 SIRNA Assays ›
Gene Symbol:
UBAP1
Gene Name:
ubiquitin associated protein 1
Gene Aliases:
NAG20, UAP, UBAP, UBAP-1
Chromosome Location:
Chr.9: 34179005 - 34252523 on Build GRCh38
UniGene ID:
Hs.268963
Species:
Human
siRNA ID
HSS181958
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: UBAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171201.1
NP_001164672.1
4, 5
1462
NM_001171202.1
NP_001164673.1
4, 5
1378
NM_001171203.2
NP_001164674.1
6, 7
1658
NM_001171204.2
NP_001164675.1
6, 7
1626
NM_016525.4
NP_057609.2
5, 6
1503
NR_033243.2
6, 7
1619
XM_006716779.3
XP_006716842.1
5, 6
1529
XM_011517898.2
XP_011516200.1
4, 5
1296
XM_011517899.1
XP_011516201.1
6, 7
1560
XM_011517900.1
XP_011516202.1
5, 6
1435
XM_017014801.1
XP_016870290.1
6, 7
1895
XM_017014802.1
XP_016870291.1
4, 5
1252
GenBank mRNA
AF222043.2
-
5, 6
1432
AK074426.1
-
5, 6
1496
AK074724.1
-
3, 4
1241
AK074745.1
-
5, 6
1463
AK074812.1
-
5, 6
1503
AK074969.1
-
6, 7
1605
AK074995.1
-
5, 6
1467
AK223235.1
-
5, 6
1458
AK295482.1
-
4, 5
1338
AK303711.1
-
4, 5
1462
AK309329.1
-
4, 5
1333
AL136733.1
-
5, 6
1296
BC020950.1
-
5, 6
1465
3851813
BC098141.1
-
5, 6
1399
40011247
BC098316.1
-
5, 6
1399
40011248
BC099726.1
-
5, 6
1399
40011249
BC100668.1
-
5, 6
1399
40011243
CR533551.1
-
4, 5
1260
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein transport
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
protein binding
ubiquitin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0