Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278710.1 | NP_001265639.1 | 3 | 246 | ||
NM_001278711.1 | NP_001265640.1 | 3 | 485 | |||
NM_016588.2 | NP_057672.1 | 2 | 744 | |||
XM_006715106.3 | XP_006715169.1 | 4 | 1135 | |||
XM_006715107.3 | XP_006715170.1 | 2 | 925 | |||
GenBank mRNA | AF114833.1 | - | 2 | 275 | ||
AF136631.1 | - | 2 | 261 | |||
AJ420483.1 | - | 2 | 218 | |||
AK093824.1 | - | 2 | 939 | |||
AK314095.1 | - | 2 | 297 | |||
AL541038.3 | - | 2 | 330 | |||
BC002683.2 | - | 2 | 222 | 3605775 | ||
BC042019.1 | - | 2 | 745 | 5312094 | ||
BF970432.1 | - | 2 | 137 | |||
BX340783.2 | - | 3 | 483 | |||
DB013243.1 | - | 3 | 246 |