Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127889.4 | NP_001121361.1 | 8 | 842 | ||
NM_001127890.4 | NP_001121362.1 | 7 | 942 | |||
NM_016630.6 | NP_057714.1 | 8 | 1023 | |||
XM_005254437.4 | XP_005254494.1 | 9 | 1056 | |||
XM_017022297.1 | XP_016877786.1 | 8 | 1391 | |||
XM_017022298.1 | XP_016877787.1 | 9 | 1481 | |||
GenBank mRNA | AF208861.1 | - | 7 | 738 | ||
AF212231.1 | - | 8 | 810 | |||
AK123725.1 | - | 6 | 1255 | |||
AK172849.1 | - | 8 | 855 | |||
AK225527.1 | - | 8 | 855 | |||
AK301362.1 | - | 7 | 907 | |||
AK312943.1 | - | 8 | 858 | |||
BC000244.1 | - | 8 | 893 | 3357388 | ||
BI258560.1 | - | 2 | 85 |