Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016647.2 | NP_057731.1 | 2 | 692 | ||
XM_005250955.2 | XP_005251012.1 | 2 | 533 | |||
GenBank mRNA | AF242773.1 | - | 2 | 625 | ||
AK055140.1 | - | 1 | 837 | |||
AK075407.1 | - | 2 | 620 | |||
AK301564.1 | - | 2 | 486 | |||
AK315614.1 | - | 2 | 643 | |||
BC001311.1 | - | 2 | 692 | 3453935 | ||
BC019104.2 | - | 2 | 624 | 5021156 |