Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242923.1 | NP_001229852.1 | 3 | 279 | ||
NM_016091.3 | NP_057175.1 | 3 | 279 | |||
XM_005261625.1 | XP_005261682.1 | 3 | 234 | |||
XM_006724260.3 | XP_006724323.1 | 3 | 877 | |||
XM_017028814.1 | XP_016884303.1 | 3 | 234 | |||
GenBank mRNA | AF077207.1 | - | 3 | 215 | ||
AF083243.1 | - | 3 | 234 | |||
AF109359.1 | - | 3 | 216 | |||
AK056129.1 | - | 3 | 783 | |||
AK093540.1 | - | 2 | 703 | |||
AK129648.1 | - | 3 | 232 | |||
AK222496.1 | - | 3 | 234 | |||
AK222529.1 | - | 3 | 232 | |||
AK293629.1 | - | 3 | 234 | |||
AK298776.1 | - | 3 | 210 | |||
AK301323.1 | - | 3 | 233 | |||
AK315533.1 | - | 3 | 240 | |||
AL357455.1 | - | 3 | 207 | |||
BC001101.2 | - | 3 | 217 | 3507964 | ||
BC007510.2 | - | 3 | 217 | 2959747 | ||
BC029265.1 | - | 3 | 219 | 5110941 | ||
CR456436.1 | - | 3 | 340 | |||
DA051005.1 | - | 3 | 279 | |||
DA663575.1 | - | 3 | 240 |