Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014306.4 | NP_055121.1 | 8 | 1005 | ||
GenBank mRNA | AF155658.1 | - | 8 | 953 | ||
AK297826.1 | - | 8 | 882 | |||
AK301945.1 | - | 8 | 913 | |||
AK312503.1 | - | 8 | 965 | |||
AL050255.1 | - | 8 | 949 | |||
BC000151.1 | - | 8 | 943 | 2984824 | ||
BC002970.1 | - | 8 | 943 | 3533883 | ||
BC010308.1 | - | 8 | 955 | 3451576 | ||
BC016707.1 | - | 8 | 970 | 4102148 | ||
CR456450.1 | - | 8 | 920 | |||
CR457175.1 | - | 8 | 874 |