Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165258.1 | NP_001158730.1 | 5 | 625 | ||
NM_016462.3 | NP_057546.1 | 5 | 611 | |||
GenBank mRNA | AF151028.1 | - | 5 | 421 | ||
AK223462.1 | - | 5 | 309 | |||
AK309591.1 | - | 5 | 359 | |||
AK310191.1 | - | 5 | 358 | |||
AL702149.1 | - | 5 | 445 | |||
BC002496.2 | - | 5 | 346 | 3050609 | ||
BC010086.2 | - | 5 | 375 | 2907072 | ||
CX784985.1 | - | 5 | 611 |