Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000284.3 | NP_000275.1 | 8 | 947 | ||
NM_001173454.1 | NP_001166925.1 | 9 | 1061 | |||
NM_001173455.1 | NP_001166926.1 | 8 | 968 | |||
NM_001173456.1 | NP_001166927.1 | 7 | 854 | |||
XM_017029574.1 | XP_016885063.1 | 8 | 955 | |||
GenBank mRNA | AK129887.1 | - | 8 | 882 | ||
AK222740.1 | - | 8 | 878 | |||
AK222988.1 | - | 8 | 912 | |||
AK293250.1 | - | 9 | 1026 | |||
AK296341.1 | - | 8 | 934 | |||
AK296457.1 | - | 7 | 768 | |||
AK312263.1 | - | 8 | 913 | |||
BC002406.1 | - | 8 | 859 | 2961286 | ||
EF590116.1 | - | 2 | 597 | |||
EF590117.1 | - | 979 | ||||
J03503.1 | - | 8 | 874 | |||
J03575.1 | - | 8 | 847 | |||
L13318.1 | - | 8 | 907 | |||
L48690.1 | - | 8 | 907 | |||
M24848.1 | - | 8 | 845 | |||
X52709.1 | - | 8 | 856 | |||
X52710.1 | - | 8 | 858 |