Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARAF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284239.1 | NP_001271168.1 | 1 | 307 | ||
NM_016127.5 | NP_057211.4 | 1 | 307 | |||
XM_017013567.1 | XP_016869056.1 | 1 | 557 | |||
XM_017013570.1 | XP_016869059.1 | 1 | 246 | |||
GenBank mRNA | AB028926.1 | - | 1 | 167 | ||
AF078855.1 | - | 1 | 136 | |||
AF100748.1 | - | 1 | 64 | |||
AF490252.1 | - | 1 | 48 | |||
AK025927.1 | - | 1 | 217 | |||
AK075340.1 | - | 1 | 165 | |||
AK094386.1 | - | 1 | 242 | |||
AK222632.1 | - | 1 | 64 | |||
AK314659.1 | - | 1 | 190 | |||
AK315956.1 | - | 1 | 64 | |||
AY359104.1 | - | 1 | 232 | |||
AY454545.1 | - | 1 | 63 | |||
BC015012.1 | - | 1 | 136 | 3908758 | ||
DC359554.1 | - | 1 | 308 | |||
JQ348891.1 | - | 1 | 176 | |||
JQ348892.1 | - | 1 | 47 |