Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202429.1 | NP_001189358.1 | 3 | 651 | ||
NM_016150.4 | NP_057234.2 | 1 | 626 | |||
XM_005267758.3 | XP_005267815.1 | 4 | 711 | |||
XM_011536834.2 | XP_011535136.1 | 3 | 717 | |||
GenBank mRNA | AB056723.1 | - | 1 | 626 | ||
AB488462.1 | - | 3 | 651 | |||
AF159164.1 | - | 1 | 14 | |||
AJ251238.1 | - | 1 | 280 | |||
AK057078.1 | - | 4 | 599 | |||
AK303686.1 | - | 3 | 470 | |||
AK315628.1 | - | 1 | 252 | |||
AL137735.1 | - | 1 | 47 | |||
BC032354.1 | - | 1 | 259 | 5218997 |