Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282150.1 | NP_001269079.1 | 4 | 292 | ||
NM_001282151.1 | NP_001269080.1 | 2 | 137 | |||
NM_016226.4 | NP_057310.1 | 2 | 137 | |||
NM_057180.2 | NP_476528.1 | 3 | 149 | |||
NR_104099.1 | 4 | 189 | ||||
NR_104100.1 | 3 | 177 | ||||
NR_104101.1 | 3 | 227 | ||||
XM_006719459.3 | XP_006719522.1 | 4 | 198 | |||
XM_006719460.3 | XP_006719523.1 | 5 | 235 | |||
XM_011538484.2 | XP_011536786.1 | 3 | 183 | |||
XM_017019457.1 | XP_016874946.1 | 3 | 208 | |||
XM_017019458.1 | XP_016874947.1 | 2 | 328 | |||
XM_017019459.1 | XP_016874948.1 | 3 | 226 | |||
GenBank mRNA | AB578891.1 | - | 3 | 118 | ||
AB578892.1 | - | 3 | 149 | |||
AB618733.1 | - | 2 | 120 | |||
AF086379.1 | - | 1 | 43 | |||
AF168716.1 | - | 3 | 107 | |||
AF175264.1 | - | 2 | 106 | |||
AF193795.1 | - | 2 | 99 | |||
AF201936.1 | - | 3 | 194 | |||
AF201946.1 | - | 2 | 85 | |||
AK129767.1 | - | 2 | 99 | |||
AK130510.1 | - | 935 | ||||
AL136614.1 | - | 2 | 115 | |||
AL832866.1 | - | 2 | 2130 | |||
BC000880.1 | - | 3 | 107 | 3461977 | ||
BC015095.1 | - | 3 | 116 | |||
BC095446.1 | - | 2 | 108 | 30719438 | ||
BF030144.1 | - | 148 | ||||
BG707325.1 | - | 2 | 132 | |||
BM806646.1 | - | 4 | 245 | |||
BU568689.1 | - | 4 | 150 | |||
CB158821.1 | - | 2 | 71 | |||
CR457182.1 | - | 1 | 60 | |||
CR533468.1 | - | 1 | 48 |