Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WWOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291997.1 | NP_001278926.1 | 1 | 453 | ||
NM_016373.3 | NP_057457.1 | 1 | 453 | |||
NM_130791.3 | NP_570607.1 | 1 | 453 | |||
NR_120435.1 | 1 | 453 | ||||
NR_120436.1 | 1 | 453 | ||||
XM_011523101.2 | XP_011521403.1 | 1 | 212 | |||
XM_011523103.2 | XP_011521405.1 | 1 | 212 | |||
XM_011523104.2 | XP_011521406.1 | 1 | 212 | |||
XM_011523105.2 | XP_011521407.1 | 1 | 212 | |||
XM_017023278.1 | XP_016878767.1 | 1 | 212 | |||
GenBank mRNA | AF187015.1 | - | 1 | 171 | ||
AF211943.1 | - | 1 | 212 | |||
AF227526.1 | - | 1 | 172 | |||
AF227529.1 | - | 1 | 191 | |||
AF395123.1 | - | 1 | 220 | |||
AF395124.1 | - | 1 | 220 | |||
AK027626.1 | - | 1 | 211 | |||
AK290438.1 | - | 1 | 203 | |||
AK291300.1 | - | 1 | 210 | |||
AK298322.1 | - | 1 | 240 | |||
AW874692.1 | - | 1 | 98 | |||
AW874693.1 | - | 1 | 432 | |||
AY256821.1 | - | 1 | 142 | |||
BC003184.1 | - | 1 | 166 | 3535809 | ||
BT007445.1 | - | 87 | ||||
BU157392.1 | - | 1 | 80 | |||
DB035405.1 | - | 1 | 453 |