Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016530.2 | NP_057614.1 | 5, 6 | 515 | ||
XM_017022312.1 | XP_016877801.1 | 6, 7 | 570 | |||
GenBank mRNA | AB038995.1 | - | 5, 6 | 510 | ||
AK001111.1 | - | 5, 6 | 438 | |||
AK297598.1 | - | 5, 6 | 438 | |||
AL833365.1 | - | 5, 6 | 428 | |||
BC020654.1 | - | 5, 6 | 427 | 4701429 |