Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000295.4 | NP_000286.3 | 3 | 1089 | ||
NM_001002235.2 | NP_001002235.1 | 3 | 1068 | |||
NM_001002236.2 | NP_001002236.1 | 5 | 1382 | |||
NM_001127700.1 | NP_001121172.1 | 3 | 1105 | |||
NM_001127701.1 | NP_001121173.1 | 5 | 1401 | |||
NM_001127702.1 | NP_001121174.1 | 4 | 1209 | |||
NM_001127703.1 | NP_001121175.1 | 5 | 1364 | |||
NM_001127704.1 | NP_001121176.1 | 5 | 1361 | |||
NM_001127705.1 | NP_001121177.1 | 5 | 1379 | |||
NM_001127706.1 | NP_001121178.1 | 4 | 1172 | |||
NM_001127707.1 | NP_001121179.1 | 4 | 1169 | |||
XM_017021370.1 | XP_016876859.1 | 4 | 1269 | |||
GenBank mRNA | AK026174.1 | - | 3 | 871 | ||
AK315637.1 | - | 4 | 1125 | |||
BC011991.1 | - | 3 | 1060 | 3859644 | ||
BC015642.2 | - | 3 | 855 | 4644658 | ||
BC070163.1 | - | 4 | 795 | |||
BT019455.1 | - | 2 | 828 | |||
BX161449.1 | - | 3 | 995 | |||
BX247968.1 | - | 5 | 1309 | |||
BX248002.1 | - | 2 | 837 | |||
BX248257.1 | - | 3 | 862 | |||
GU727620.1 | - | 2 | 829 | |||
K01396.1 | - | 2 | 847 | |||
KU178257.1 | - | 2 | 828 | |||
X01683.1 | - | 2 | 866 | |||
X17122.1 | - | 2 | 407 |