Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039366.1 | NP_001034455.1 | 1 | 201 | ||
NM_001039367.1 | NP_001034456.1 | 1 | 201 | |||
NM_001696.3 | NP_001687.1 | 1 | 201 | |||
GenBank mRNA | AK294623.1 | - | 1 | 128 | ||
AK312394.1 | - | 1 | 122 | |||
AK315941.1 | - | 1 | 127 | |||
BC004443.1 | - | 1 | 103 | 2820930 | ||
BE735148.1 | - | 1 | 147 | |||
BI546032.1 | - | 1 | 153 | |||
BI597419.1 | - | 1 | 131 | |||
BQ888820.1 | - | 1 | 146 | |||
BT007128.1 | - | 1 | 14 | |||
CK005516.1 | - | 1 | 204 | |||
CR456385.1 | - | 1 | 67 | |||
X71491.1 | - | 1 | 70 | |||
X76228.1 | - | 1 | 89 |