Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000296.3 | NP_000287.3 | 15 | 3522 | ||
NM_001009944.2 | NP_001009944.2 | 15 | 3522 | |||
XM_005255370.3 | XP_005255427.1 | 5 | 1109 | |||
XM_011522528.2 | XP_011520830.1 | 16 | 3547 | |||
XM_011522529.2 | XP_011520831.1 | 16 | 3548 | |||
XM_011522530.2 | XP_011520832.1 | 16 | 3352 | |||
XM_011522532.2 | XP_011520834.1 | 15 | 3280 | |||
XM_011522534.2 | XP_011520836.1 | 16 | 3337 | |||
XM_011522535.2 | XP_011520837.1 | 7 | 1569 | |||
XM_011522537.2 | XP_011520839.1 | 5 | 480 | |||
XM_017023285.1 | XP_016878774.1 | 6 | 1124 | |||
GenBank mRNA | L33243.1 | - | 15 | 3524 | ||
U24497.1 | - | 15 | 3448 |