Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039840.2 | NP_001034929.2 | 3 | 614 | ||
NM_001300884.1 | NP_001287813.1 | 3 | 614 | |||
XM_017029582.1 | XP_016885071.1 | 3 | 717 | |||
XM_017029583.1 | XP_016885072.1 | 4 | 295 | |||
GenBank mRNA | AK295903.1 | - | 3 | 617 | ||
AK297768.1 | - | 3 | 471 | |||
AL832095.1 | - | 3 | 254 | |||
BC127727.1 | - | 3 | 525 | 40127524 | ||
BC127728.1 | - | 3 | 525 | 40127525 | ||
BC136691.1 | - | 3 | 519 | 9020681 | ||
BC136692.1 | - | 3 | 519 | 9020682 | ||
CR936642.1 | - | 3 | 565 |