Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S1PR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166215.1 | NP_001159687.1 | 2 | 334 | ||
NM_030760.4 | NP_110387.1 | 2 | 218 | |||
GenBank mRNA | AF331840.1 | - | 1 | 156 | ||
AF331841.1 | - | 1 | 157 | |||
AF331842.1 | - | 1 | 157 | |||
AK074661.1 | - | 2 | 273 | |||
AK094880.1 | - | 2 | 216 | |||
BC034703.1 | - | 2 | 204 | 4754850 | ||
BC067781.1 | - | 2 | 219 | 5286249 | ||
DA747912.1 | - | 2 | 219 | |||
DA866378.1 | - | 2 | 284 | |||
DB013424.1 | - | 2 | 338 |
Set Membership: |
G-protein coupled receptor |