Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317009.1 | NP_001303938.1 | 6 | 2070 | ||
NM_053277.2 | NP_444507.1 | 5 | 2016 | |||
XM_017028406.1 | XP_016883895.1 | 5 | 2226 | |||
GenBank mRNA | AF426169.1 | - | 2 | 147 | ||
AF448438.1 | - | 5 | 1790 | |||
AF448439.1 | - | 6 | 1844 | |||
AK092733.1 | - | 4 | 331 | |||
AK289663.1 | - | 5 | 2016 | |||
BC040196.2 | - | 5 | 956 |