Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127218.2 | NP_001120690.1 | 2 | 437 | ||
NM_001256879.1 | NP_001243808.1 | 3 | 798 | |||
NM_006230.3 | NP_006221.2 | 2 | 264 | |||
XM_011515431.1 | XP_011513733.1 | 2 | 260 | |||
GenBank mRNA | AK292347.1 | - | 3 | 798 | ||
AK312294.1 | - | 2 | 397 | |||
AU117477.2 | - | 2 | 242 | |||
BC000459.2 | - | 2 | 227 | 2822169 | ||
BC030001.2 | - | 1 | 204 | |||
BP224472.1 | - | 2 | 264 | |||
DB025069.1 | - | 2 | 496 | |||
U21090.1 | - | 2 | 226 |