Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018973.3 | NP_061846.2 | 1 | 408 | ||
NM_153741.1 | NP_714963.1 | 2 | 276 | |||
XM_017001498.1 | XP_016856987.1 | 2 | 355 | |||
GenBank mRNA | AB028128.1 | - | 1 | 258 | ||
AF312922.1 | - | 1 | 408 | |||
AF312923.1 | - | 1 | 258 | |||
AK293625.1 | - | 1 | 532 | |||
BC032223.2 | - | 1 | 247 | |||
BC065233.1 | - | 2 | 284 | |||
BC104202.1 | - | 1 | 381 | 40031374 | ||
BC104203.2 | - | 1 | 381 | 40031376 | ||
BC104481.1 | - | 2 | 304 |